Learn More
Medchemexpress LLC Maltose monohydrate | 6363-53-7 | 360.31 | 1 ML
SDP
Catalog No. 5000414685
Encompass_Preferred
Maltose monohydrate is a degradative product of Starch and a substrate of α-Glucosidase. α-Glucosidase hydrolyzes Maltose to Glucose. Maltose monohydrate can protect proteins, membranes, and the photosynthetic electron transport chain under heat stress and freezing stress.
  • Degradative product of starch
  • Substrate of α-Glucosidase
  • Protects proteins, membranes, and the photosynthetic electron transport chain under heat stress and freezing stress
Catalog No. 50-004-14685 Supplier Medchemexpress LLC Supplier No. HYN2024A10MM/1ML
Maltose monohydrate is a degradative product of Starch and a substrate of α-Glucosidase. α-Glucosidase hydrolyzes Maltose to Glucose. Maltose monohydrate can protect proteins, membranes, and the photosynthetic electron transport chain under heat stress and freezing stress.
  • Degradative product of starch
  • Substrate of α-Glucosidase
  • Protects proteins, membranes, and the photosynthetic electron transport chain under heat stress and freezing stress