Learn More
Medchemexpress LLC Maltose monohydrate | 6363-53-7 | 360.31 | 25 MG
SDP
Catalog No. 5000389740
Encompass_Preferred
Maltose monohydrate is a degradative product of starch and a substrate of α-Glucosidase, which hydrolyses maltose to glucose. This compound protects proteins, membranes, and the photosynthetic electron transport chain under heat and freezing stress. It also serves as an energy source for bacteria and its levels increase in leaves during starch breakdown under photorespiratory conditions. As an analytical standard, it is intended for research and analytical applications.
  • Protects proteins, membranes, and the photosynthetic electron transport chain under heat stress and freezing stress.
  • Serves as an analytical standard and reference in assays.
  • Employed in qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
Catalog No. 50-003-89740 Supplier Medchemexpress LLC Supplier No. 25MGHYN2024AR
Maltose monohydrate is a degradative product of starch and a substrate of α-Glucosidase, which hydrolyses maltose to glucose. This compound protects proteins, membranes, and the photosynthetic electron transport chain under heat and freezing stress. It also serves as an energy source for bacteria and its levels increase in leaves during starch breakdown under photorespiratory conditions. As an analytical standard, it is intended for research and analytical applications.
  • Protects proteins, membranes, and the photosynthetic electron transport chain under heat stress and freezing stress.
  • Serves as an analytical standard and reference in assays.
  • Employed in qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.