Learn More
Medchemexpress LLC Maltose monohydrate | 6363-53-7 | 360.31 | 50 MG
SDP
Catalog No. 5000415939
Encompass_Preferred
Maltose monohydrate (Standard) is an analytical standard intended for research and analytical applications. It serves as an energy source for bacteria and is known to protect proteins, membranes, and the photosynthetic electron transport chain.
  • Analytical standard
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Energy source for bacteria
  • Protects proteins, membranes, and photosynthetic electron transport chain
Catalog No. 50-004-15939 Supplier Medchemexpress LLC Supplier No. HYN2024AR50MG
Maltose monohydrate (Standard) is an analytical standard intended for research and analytical applications. It serves as an energy source for bacteria and is known to protect proteins, membranes, and the photosynthetic electron transport chain.
  • Analytical standard
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Energy source for bacteria
  • Protects proteins, membranes, and photosynthetic electron transport chain