Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maltose monohydrate-d14 | 6363-53-7 | 93.7% | 374.40 | C12H10D14O12 | 10 MG
SDP

Catalog No. 5000295581
Encompass_Preferred

Maltose monohydrate-d14 is the deuterium-labeled form of maltose monohydrate used as a stable-isotope tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. It is intended for research use in metabolic and pharmacokinetic studies.

  • Deuterium-labeled tracer for analytical quantification.
  • Useful as an internal standard in NMR, GC-MS, and LC-MS assays.
  • Purity approximately 93.7%.
  • Molecular weight about 374.40 g/mol.
  • White to off-white solid, soluble in water (≈100 mg/mL; ultrasonic and warming may be required).
  • Recommended storage sealed and away from moisture; in solvent: -80 °C (6 months) or -20 °C (1 month).

Catalog No. 50-002-95581 Supplier Medchemexpress LLC Supplier No. HYN2024AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maltose monohydrate-d14 is the deuterium-labeled form of maltose monohydrate used as a stable-isotope tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. It is intended for research use in metabolic and pharmacokinetic studies.

  • Deuterium-labeled tracer for analytical quantification.
  • Useful as an internal standard in NMR, GC-MS, and LC-MS assays.
  • Purity approximately 93.7%.
  • Molecular weight about 374.40 g/mol.
  • White to off-white solid, soluble in water (≈100 mg/mL; ultrasonic and warming may be required).
  • Recommended storage sealed and away from moisture; in solvent: -80 °C (6 months) or -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.