Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose, Monohydrate, Powder,, MilliporeSigma™

Catalog No. MX016020 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MX016020 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MX016020 Supplier MilliporeSigma Supplier No. MX016020

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Color Tan-White
Quantity 10 kg
Formula Weight 360.32
Physical Form Powder
Chemical Name or Material Maltose, Monohydrate, Powder
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.