Learn More
Medchemexpress LLC Maltose monohydrate (Standard) | 6363-53-7 | 360.31 | 100 MG
SDP
Catalog No. 5000415923
Encompass_Preferred
Maltose monohydrate (Standard) is the analytical standard of Maltose monohydrate, intended for research and analytical applications. It serves as an energy source for bacteria and its levels increase in leaves during starch breakdown under photorespiratory conditions. It is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Serves as an energy source for bacteria
  • Protects proteins, membranes, and the photosynthetic electron transport chain
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-15923 Supplier Medchemexpress LLC Supplier No. HYN2024AR100MG
Maltose monohydrate (Standard) is the analytical standard of Maltose monohydrate, intended for research and analytical applications. It serves as an energy source for bacteria and its levels increase in leaves during starch breakdown under photorespiratory conditions. It is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Serves as an energy source for bacteria
  • Protects proteins, membranes, and the photosynthetic electron transport chain
  • Used in HPLC, GC, and MS research experiments