Learn More
Medchemexpress LLC Maltose phosphorylase | 9030-19-7 | 250 U
SDP
Catalog No. 5000396213
Encompass_Preferred
Maltose phosphorylase is a dimerase that catalyzes the transformation of maltose and inorganic phosphate into β-D-glucose-1-phosphate and glucose. It is classified in family 65 of the glycoside hydrolases.
  • Catalyzes transformation of maltose and inorganic phosphate
  • Classified in family 65 of glycoside hydrolases
  • Enzyme activity: ≥9 U/mg solid
  • Specific activity: ≥100 U/mg protein
Catalog No. 50-003-96213 Supplier Medchemexpress LLC Supplier No. 250UHYP2741
Maltose phosphorylase is a dimerase that catalyzes the transformation of maltose and inorganic phosphate into β-D-glucose-1-phosphate and glucose. It is classified in family 65 of the glycoside hydrolases.
  • Catalyzes transformation of maltose and inorganic phosphate
  • Classified in family 65 of glycoside hydrolases
  • Enzyme activity: ≥9 U/mg solid
  • Specific activity: ≥100 U/mg protein