Learn More
Medchemexpress LLC Maltose phosphorylase | 9030-19-7 | 50 U
SDP
Catalog No. 5000398677
Encompass_Preferred
Maltose phosphorylase is a dimerase enzyme that catalyzes the transformation of maltose and inorganic phosphate into β-D-glucose-1-phosphate and glucose. This enzyme is classified within family 65 of the glycoside hydrolases.
  • Catalyzes maltose transformation
  • Forms β-D-glucose-1-phosphate and glucose
  • Classified in family 65 of glycoside hydrolases
Catalog No. 50-003-98677 Supplier Medchemexpress LLC Supplier No. HYP274150U
Maltose phosphorylase is a dimerase enzyme that catalyzes the transformation of maltose and inorganic phosphate into β-D-glucose-1-phosphate and glucose. This enzyme is classified within family 65 of the glycoside hydrolases.
  • Catalyzes maltose transformation
  • Forms β-D-glucose-1-phosphate and glucose
  • Classified in family 65 of glycoside hydrolases