Learn More
Bioworld MALTOSE SEPAROPORE 4B
Catalog No. 50197637
Encompass
Maltose coupled with Separopore 4B. Specifications: Ligand: Maltose.; Particle size (range): 50-175m.; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B (Agarose 4% Beads).; pH stability: 4-9.; Flow specifications: 70-140 cm/h...
Catalog No. 50-197-637 Supplier Bioworld Supplier No. 201700191
Maltose coupled with Separopore 4B. Specifications: Ligand: Maltose.; Particle size (range): 50-175m.; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B (Agarose 4% Beads).; pH stability: 4-9.; Flow specifications: 70-140 cm/h...