Learn More
Bioworld MALTOSE SEPAROPOREAGAROSE
Catalog No. 50197852
Encompass
MALTOSE SEPAROPORE (AGAROSE) 4B: Maltose coupled with Separopore 4B. Specifications: Ligand: Maltose.; Particle size (range): 50-175m.; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B (Agarose 4% Beads).; pH stability: 4-9.;...
Catalog No. 50-197-852 Supplier Bioworld Supplier No. 201810721
MALTOSE SEPAROPORE (AGAROSE) 4B: Maltose coupled with Separopore 4B. Specifications: Ligand: Maltose.; Particle size (range): 50-175m.; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B (Agarose 4% Beads).; pH stability: 4-9.;...