Learn More
Medchemexpress LLC Maltose (standard) | 69-79-4 | 342.30 | 100 MG
SDP
Catalog No. 5000415924
Encompass_Preferred
Maltose (standard) is the analytical standard of Maltose. This product is intended for research and analytical applications. Maltose is a disaccharide formed from two units of glucose joined with an α(1→4) bond, a reducing sugar. Maltose monohydrate can be used as an energy source for bacteria.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research use only.
  • Reference standard supplied assay.
Catalog No. 50-004-15924 Supplier Medchemexpress LLC Supplier No. HYN2024R100MG
Maltose (standard) is the analytical standard of Maltose. This product is intended for research and analytical applications. Maltose is a disaccharide formed from two units of glucose joined with an α(1→4) bond, a reducing sugar. Maltose monohydrate can be used as an energy source for bacteria.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research use only.
  • Reference standard supplied assay.