Learn More
Medchemexpress LLC Maltose (Standard) | 69-79-4 | 342.30 | 25 MG
SDP
Catalog No. 5000390075
Encompass_Preferred
Maltose (Standard) is an analytical standard used for research and analytical applications. This disaccharide, composed of two glucose units linked by an α(1→4) bond, is a reducing sugar. Maltose monohydrate can also provide an energy source for bacteria.
  • Analytical standard for research and analytical applications
  • Disaccharide formed from two units of glucose
  • Classified as a reducing sugar
  • Can function as an energy source for bacteria
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-003-90075 Supplier Medchemexpress LLC Supplier No. 25MGHYN2024R
Maltose (Standard) is an analytical standard used for research and analytical applications. This disaccharide, composed of two glucose units linked by an α(1→4) bond, is a reducing sugar. Maltose monohydrate can also provide an energy source for bacteria.
  • Analytical standard for research and analytical applications
  • Disaccharide formed from two units of glucose
  • Classified as a reducing sugar
  • Can function as an energy source for bacteria
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS