Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maltose (Standard) | 69-79-4 | 342.30 | 250 MG
SDP

Catalog No. 5000389456
Encompass_Preferred

Maltose (Standard) is the analytical standard of Maltose, intended for research and analytical applications. It is a disaccharide formed from two units of glucose joined with an α(1→4) bond, a reducing sugar. This compound is the grade of analytical standard, which is the reference standard supplied assay.

  • Intended for research and analytical applications.
  • Disaccharide formed from two units of glucose.
  • Can be used as an energy source for bacteria.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Purity: 99.81%.
  • Appearance: solid, white to off-white.
  • Initial source: endogenous metabolite.

Catalog No. 50-003-89456 Supplier Medchemexpress LLC Supplier No. 250MGHYN2024R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maltose (Standard) is the analytical standard of Maltose, intended for research and analytical applications. It is a disaccharide formed from two units of glucose joined with an α(1→4) bond, a reducing sugar. This compound is the grade of analytical standard, which is the reference standard supplied assay.

  • Intended for research and analytical applications.
  • Disaccharide formed from two units of glucose.
  • Can be used as an energy source for bacteria.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Purity: 99.81%.
  • Appearance: solid, white to off-white.
  • Initial source: endogenous metabolite.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.