Learn More
Medchemexpress LLC Maltose (Standard) | 69-79-4 | 342.30 | 50 MG
SDP
Catalog No. 5000415167
Encompass_Preferred
Maltose (Standard) is an analytical standard of Maltose, intended for research and analytical applications. This disaccharide is formed from two units of glucose joined with an α(1→4) bond and is a reducing sugar. Maltose monohydrate can be used as an energy source for bacteria and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Disaccharide formed from two units of glucose
  • Functions as a reducing sugar
  • Can serve as an energy source for bacteria
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-15167 Supplier Medchemexpress LLC Supplier No. HYN2024R50MG
Maltose (Standard) is an analytical standard of Maltose, intended for research and analytical applications. This disaccharide is formed from two units of glucose joined with an α(1→4) bond and is a reducing sugar. Maltose monohydrate can be used as an energy source for bacteria and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Disaccharide formed from two units of glucose
  • Functions as a reducing sugar
  • Can serve as an energy source for bacteria
  • Suitable for HPLC, GC, and MS research experiments