Learn More
Cayman Chemical Maltotetraose 50mg
SDP
Catalog No. 502935666
Encompass_Preferred
A tetrasaccharide; increases the α-amylase synthesis rate in B. stearothermophilus three-fold greater than sucrose or glucose when used at at 0.1 mM; inhibits the growth of E. carotovora in a cylinder-agar plate assay at 750 μg/ml; inhibits TNF-α-induced expression of ICAM-1 in MOVAS-1 mouse VSMCs
Catalog No. 50-293-5666 Supplier Cayman Chemical Supplier No. 2497550 MG
A tetrasaccharide; increases the α-amylase synthesis rate in B. stearothermophilus three-fold greater than sucrose or glucose when used at at 0.1 mM; inhibits the growth of E. carotovora in a cylinder-agar plate assay at 750 μg/ml; inhibits TNF-α-induced expression of ICAM-1 in MOVAS-1 mouse VSMCs