Learn More
Medchemexpress LLC HY-N2464 5mg Medchemexpress, Maltotetraose CAS:34612-38-9 Purity:>98%
SDP
Catalog No. 501933928
Encompass_Preferred
Medchemexpress, HY-N2464 5mg Maltotetraose CAS:34612-38-9 Maltotetraose can be used as a substrate for the enzyme-coupled determination of amylase activity in biological fluids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3928 Supplier Medchemexpress LLC Supplier No. HYN24645MG
Medchemexpress, HY-N2464 5mg Maltotetraose CAS:34612-38-9 Maltotetraose can be used as a substrate for the enzyme-coupled determination of amylase activity in biological fluids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.