Learn More
Carbosynth LLC. Maltotetraose, 34612-38-9, MFCD00010576, 25mg
SDP
Supplier:  Carbosynth LLC. OM0279625MG
Encompass_Preferred
Chemical Formula: C24H42O21, MW: 666.58, Synonyms: a-D-Glucopyranosyl-1,4-O-a-D-glucopyranosyl-1,4-O-a-D-glucopyranosyl-1,4-D-glucopyranose; Amylotetraose.
Catalog No. NC1482451