Learn More
U.S. Pharmacopeia Maltotriose, 1109-28-0, MFCD00006629, 500 mg
Catalog No. NC1038156
Encompass
Empirical Formula (Hill Notation): C18H32O16, Molecular Weight: 504.44, Synonym: α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-Glc, Maltotriose, O-α-DD-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose.
Catalog No. NC1038156 Supplier U.S. Pharmacopeia Supplier No. 1375047
Empirical Formula (Hill Notation): C18H32O16, Molecular Weight: 504.44, Synonym: α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-Glc, Maltotriose, O-α-DD-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose.