Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maltulose monohydrate | 207511-09-9 | MFCD00217057 | 99.8% | 360.31 g/mol | C12H24O12 | 100 MG
SDP

Catalog No. 5000281159
Encompass_Preferred

Maltulose monohydrate is a research-grade disaccharide (4-O-α-D-glucopyranosyl-D-fructose monohydrate) used as an energy source for bacteria and as a biomaterial in life-science research. Supplied as a high-purity solid for laboratory applications, it is intended for research use only.

  • Chemical formula C12H24O12.
  • Molecular weight 360.31 g/mol.
  • CAS number 207511-09-9.
  • Purity 99.8%.
  • Soluble in water (≈175 mg/mL); ultrasonic may be required.
  • Appearance white to off-white solid.
  • Store sealed, away from moisture; long-term storage at -20°C.
  • Intended for research use only, not for clinical or food use.

Catalog No. 50-002-81159 Supplier Medchemexpress LLC Supplier No. HYW415909100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maltulose monohydrate is a research-grade disaccharide (4-O-α-D-glucopyranosyl-D-fructose monohydrate) used as an energy source for bacteria and as a biomaterial in life-science research. Supplied as a high-purity solid for laboratory applications, it is intended for research use only.

  • Chemical formula C12H24O12.
  • Molecular weight 360.31 g/mol.
  • CAS number 207511-09-9.
  • Purity 99.8%.
  • Soluble in water (≈175 mg/mL); ultrasonic may be required.
  • Appearance white to off-white solid.
  • Store sealed, away from moisture; long-term storage at -20°C.
  • Intended for research use only, not for clinical or food use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.