Learn More
Cayman Chemical Rubusoside, Purity: ≥98%, CAS: 64849-39-4, 50 mg
SDP
Catalog No. 502940475
Encompass_Preferred
A natural non-caloric sweetener; inhibits glucose transport by Glut 1 and fructose transport by Glut5 (IC50s = 4.6 and 6.7 mM, respectively, for the recombinant human transporters); prevents palmitic acid-induced lipotoxicity in INS-1 mouse insulinoma cells at 50-200 µM; has been used to enhance the solubility of etoposide
Catalog No. 50-294-0475 Supplier Cayman Chemical Supplier No. 2904050 MG
A natural non-caloric sweetener; inhibits glucose transport by Glut 1 and fructose transport by Glut5 (IC50s = 4.6 and 6.7 mM, respectively, for the recombinant human transporters); prevents palmitic acid-induced lipotoxicity in INS-1 mouse insulinoma cells at 50-200 µM; has been used to enhance the solubility of etoposide