Learn More
Medchemexpress LLC Malvidin-3-galactoside (chloride) | 30113-37-2 | 1 MG
SDP
Catalog No. 5000417850
Encompass_Preferred
Malvidin-3-galactoside chloride is an anthocyanin monomer known for its anti-tumor functions. It induces cell cycle arrest and apoptosis in hepatocellular carcinoma (HCC) cells, and inhibits the production and accumulation of Reactive Oxygen Species (ROS).
  • Exhibits cytotoxicity on Huh-7 cells
  • Arrests cells in the S phase
  • Increases cell apoptosis in Huh-7 cells
  • Inhibits the production and accumulation of ROS
  • Modulates phosphorylation levels of ERK, p38, and JNK
  • Inhibits migration and invasion in Huh-7 cells
Catalog No. 50-004-17850 Supplier Medchemexpress LLC Supplier No. HYN66231MG
Malvidin-3-galactoside chloride is an anthocyanin monomer known for its anti-tumor functions. It induces cell cycle arrest and apoptosis in hepatocellular carcinoma (HCC) cells, and inhibits the production and accumulation of Reactive Oxygen Species (ROS).
  • Exhibits cytotoxicity on Huh-7 cells
  • Arrests cells in the S phase
  • Increases cell apoptosis in Huh-7 cells
  • Inhibits the production and accumulation of ROS
  • Modulates phosphorylation levels of ERK, p38, and JNK
  • Inhibits migration and invasion in Huh-7 cells