Learn More
Medchemexpress LLC Malvidin-3-O-arabinoside chloride | 28500-04-1 | 97.1% | 498.86 | 5 MG
SDP
Catalog No. 5000426941
Encompass_Preferred
Malvidin-3-O-arabinoside chloride (M3A chloride) is a research chemical that ameliorates ethyl carbamate-induced oxidative damage by stimulating AMPK-mediated autophagy. It is used in studies to understand cellular processes related to oxidative stress and autophagy.
  • Stimulates phosphorylation of AMP-activated protein kinase (AMPK)
  • Upregulates LAMP-1 expression
  • Restores lysosomal acidity and strengthens lysosomal functions
  • Inhibits excess reactive oxygen species (ROS) generation
  • Enhances glutathione (GSH) levels and autophagy flux
  • Upregulates LC3-II/LC3-I ratio
Catalog No. 50-004-26941 Supplier Medchemexpress LLC Supplier No. HYN93495MG
Malvidin-3-O-arabinoside chloride (M3A chloride) is a research chemical that ameliorates ethyl carbamate-induced oxidative damage by stimulating AMPK-mediated autophagy. It is used in studies to understand cellular processes related to oxidative stress and autophagy.
  • Stimulates phosphorylation of AMP-activated protein kinase (AMPK)
  • Upregulates LAMP-1 expression
  • Restores lysosomal acidity and strengthens lysosomal functions
  • Inhibits excess reactive oxygen species (ROS) generation
  • Enhances glutathione (GSH) levels and autophagy flux
  • Upregulates LC3-II/LC3-I ratio