Learn More
Cayman Chemical MalvIdInchlorIde 10mg
SDP
Catalog No. 502981627
Encompass_Preferred
An O-methylated anthocyanidin with antioxidant, antihypertensive, anti-inflammatory, and antiproliferative effects
Catalog No. 50-298-1627 Supplier Cayman Chemical Supplier No. 1975210 MG
An O-methylated anthocyanidin with antioxidant, antihypertensive, anti-inflammatory, and antiproliferative effects