Learn More
Promocell GmbH PromoCell Mammary Epithelial Cell Basal Medium 500 ml
Catalog No. 50306182
Encompass
The PromoCell Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for rat and murine mammary epithelial cells as well as mamma carcinoma cell lines.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml |
Catalog No. 50-306-182 Supplier Promocell GmbH Supplier No. C21210
The PromoCell Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for rat and murine mammary epithelial cells as well as mamma carcinoma cell lines.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.5 μg / ml |