Learn More
Cayman Chemical MAM2201 N 5chloropentyl n 10mg
SDP
Catalog No. 502955899
Encompass_Preferred
Differs from MAM2201 by having chlorine rather than fluorine on the terminal carbon of the alkyl group; intended for forensic and research applications
Catalog No. 50-295-5899 Supplier Cayman Chemical Supplier No. 1310310 MG
Differs from MAM2201 by having chlorine rather than fluorine on the terminal carbon of the alkyl group; intended for forensic and research applications