Learn More
Cayman Chemical MAM2201 NpentnoIc ACD meta
SDP
Supplier:  Cayman Chemical 14366100 UG
Encompass_Preferred
An internal standard for the quantification of MAM2201 N-pentanoic acid metabolite by GC- or LC-MS; intended for forensic and research applications
Catalog No. 50-295-3890