Learn More
Cayman Chemical MAM2201 NpentnoIc ACD meta
SDP
Catalog No. 502954117
Encompass_Preferred
An internal standard for the quantification of MAM2201 N-pentanoic acid metabolite by GC- or LC-MS; intended for forensic and research applications
Catalog No. 50-295-4117 Supplier Cayman Chemical Supplier No. 14366500 UG
An internal standard for the quantification of MAM2201 N-pentanoic acid metabolite by GC- or LC-MS; intended for forensic and research applications