Learn More
Medchemexpress LLC Mambalgin 1 TFA | 95.7% | 6554.54 | 100 UG
SDP
Catalog No. 5000422437
Encompass_Preferred
Mambalgin 1 TFA is a selective ASIC1a inhibitor with IC50 values of 192 nM for human ASIC1a and 72 nM for the ASIC1a/1b dimer. It binds to closed/inactive channels and exhibits selectivity for ASIC1a over ASIC2a, ASIC3, TRPV1, P2X2, 5-HT3, Nav1.8, Cav3.2, and Kv1.2 channels. It has been shown to increase the latency of withdrawal response in mouse tail-flick and paw-flick tests. This product is for research use only.
  • Selective ASIC1a inhibitor
  • Binds to closed/inactive channels
  • Increases latency of withdrawal response in mouse tail-flick and paw-flick tests
  • For research use only
Catalog No. 50-004-22437 Supplier Medchemexpress LLC Supplier No. HYP1441A100UG
Mambalgin 1 TFA is a selective ASIC1a inhibitor with IC50 values of 192 nM for human ASIC1a and 72 nM for the ASIC1a/1b dimer. It binds to closed/inactive channels and exhibits selectivity for ASIC1a over ASIC2a, ASIC3, TRPV1, P2X2, 5-HT3, Nav1.8, Cav3.2, and Kv1.2 channels. It has been shown to increase the latency of withdrawal response in mouse tail-flick and paw-flick tests. This product is for research use only.
  • Selective ASIC1a inhibitor
  • Binds to closed/inactive channels
  • Increases latency of withdrawal response in mouse tail-flick and paw-flick tests
  • For research use only