Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 84441MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: MAML2/1302. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a. Positive Control: MCF-7 cells. Pancreas or Placenta. Bladder or Colon Carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length MAML2. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 125kDa.Chromosome: 11q21. Gene Symbol: MAML2. Gene ID: 84441. Ensemble: ENSG00000184384. Uniprot: XP_011541325.
Catalog No. 50-188-5309