Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 55534MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: MAML3/1303. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: MCF-7 cells. Pancreas or Placenta. Cervix or Colon Carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length MAML3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 150-170kDa.Chromosome: 4q28. Gene Symbol: MAML3. Gene ID: 55534. Ensemble: ENSG00000196782. Uniprot: NP_061187.
Catalog No. 50-188-5078