Learn More
Diagnostic Biosystems MAMMAGLOBIN A Mammaglobin A
Catalog No. 502407604
Encompass_Preferred
The mammaglobin gene was first identified using a differential screening approach directed at the isolation of novel, human breast cancer-associated genes. Mammaglobin-A is highly overexpressed in breast cancer cell lines and primary breast tumors. This pattern of expression is restricted to mammary epithelium and metastatic breast tumors. Thus, mammaglobin-Aspecific T cell immune responses may provide an important approach for the design of breast cancer-specific immunotherapy.
Catalog No. 50-240-7604 Supplier Diagnostic Biosystems Supplier No. RMPD036
The mammaglobin gene was first identified using a differential screening approach directed at the isolation of novel, human breast cancer-associated genes. Mammaglobin-A is highly overexpressed in breast cancer cell lines and primary breast tumors. This pattern of expression is restricted to mammary epithelium and metastatic breast tumors. Thus, mammaglobin-Aspecific T cell immune responses may provide an important approach for the design of breast cancer-specific immunotherapy.