Learn More
Teknova Mammalian Cell lysis Buffer, 500mL, Sterile
SDP
Catalog No. 502387715
Encompass
500mM Tris Base,2.0M Sodium Chloride,5.0% Tween-20,Adjusted to pH 8.5,10X concentration,Volume provided is sufficient for 5 L of cell culture harvest
Catalog No. 50-238-7715 Supplier Teknova Supplier No. 20746
500mM Tris Base,2.0M Sodium Chloride,5.0% Tween-20,Adjusted to pH 8.5,10X concentration,Volume provided is sufficient for 5 L of cell culture harvest