Learn More
Electron Microscopy Sciences Mammalian Ringer Solution 1 LT
SDP
Catalog No. 50980246
Encompass_Preferred
Solution made up of salts including sodium chloride, potassium chloride, calcium chloride hexahydrate, sodium bicarbonate, and magnesium chloride hexahydrate.
Catalog No. 50-980-246 Supplier Electron Microscopy Sciences Supplier No. 1176310
Solution made up of salts including sodium chloride, potassium chloride, calcium chloride hexahydrate, sodium bicarbonate, and magnesium chloride hexahydrate.