Learn More
Electron Microscopy Sciences Mammalian Ringer Solution 500 ML
SDP
Catalog No. 50980245
Encompass_Preferred
Solution made up of salts including sodium chloride, potassium chloride, calcium chloride hexahydrate, sodium bicarbonate, and magnesium chloride hexahydrate.
Catalog No. 50-980-245 Supplier Electron Microscopy Sciences Supplier No. 1176305
Solution made up of salts including sodium chloride, potassium chloride, calcium chloride hexahydrate, sodium bicarbonate, and magnesium chloride hexahydrate.