Learn More
ATCC Mammaliicoccus sciuri; GV252
Catalog No. 502381100
Encompass
Mammaliicoccus sciuri strain GV252 is a whole-genome sequenced bacterium that was isolated from the skin of a southern flying squirrel. This strain has applications as a quality control strain for bioMérieux VITEK 2 products.
Catalog No. 50-238-1100 Supplier ATCC Supplier No. 29061
Mammaliicoccus sciuri strain GV252 is a whole-genome sequenced bacterium that was isolated from the skin of a southern flying squirrel. This strain has applications as a quality control strain for bioMérieux VITEK 2 products.