Learn More
ATCC Mammary Epithelial Cell Basal Medium
Catalog No. 502381821
Encompass
Mammary Epithelial Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium designed to support epithelial cells derived from a normal patient's breast. Mammary Epithelial Cell Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. To support the proliferation and plating efficiency of mammary epithelial cells ATCC PCS-600-010, Mammary Cell Basal Medium must be supplemented with the Mammary Epithelial Cell Growth Kit (ATCC PCS-600-040). ATCC’s Human Mammary Epithelial Cells cultured in Mammary Epithelial Cell complete medium (basal medium plus growth kit) is an optimal serum-free culture model for many research areas. Common uses of HMEC include the study of breast cancer development, three-dimensional culture, and carcinogen screening.
Catalog No. 50-238-1821 Supplier ATCC Supplier No. PCS600030
Mammary Epithelial Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium designed to support epithelial cells derived from a normal patient's breast. Mammary Epithelial Cell Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. To support the proliferation and plating efficiency of mammary epithelial cells ATCC PCS-600-010, Mammary Cell Basal Medium must be supplemented with the Mammary Epithelial Cell Growth Kit (ATCC PCS-600-040). ATCC’s Human Mammary Epithelial Cells cultured in Mammary Epithelial Cell complete medium (basal medium plus growth kit) is an optimal serum-free culture model for many research areas. Common uses of HMEC include the study of breast cancer development, three-dimensional culture, and carcinogen screening.