Learn More
Thomson Instrument Company Mammoth™ 30mL Automation Tips, 10/case
SDP
Catalog No. 5000887325
Encompass_Preferred
Thomson 30mL Automation Tips are high-capacity automation tips designed to bring large-volume liquid handling onto compatible liquid handling platforms With a recommended volume range of 1 5-20mL per stroke Tips help reduce manual serological pipetting and repeated low-volume aspiration/dispense cycles
Catalog No. 50-008-87325 Supplier Thomson Instrument Company Supplier No. 9453030
Thomson 30mL Automation Tips are high-capacity automation tips designed to bring large-volume liquid handling onto compatible liquid handling platforms With a recommended volume range of 1 5-20mL per stroke Tips help reduce manual serological pipetting and repeated low-volume aspiration/dispense cycles