Learn More
ACROBiosystems PE-Labeled Rhesus macaque Mamu-A*01&B2M Tetramer Protein (Peptide free)
Catalog No. 502719183
Encompass_Preferred
PE-Labeled Rhesus macaque Mamu-A*01&B2M Tetramer Protein (MAM-RP2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Pro 307 (Mamu-A*01) & Ile 21 - Met 119 (B2M) (Accession # Q5TM76 (Mamu-A*01) & Q6V7J5 (B2M)).
Catalog No. 50-271-9183 Supplier ACROBiosystems Supplier No. MAMRP2H4200UG
PE-Labeled Rhesus macaque Mamu-A*01&B2M Tetramer Protein (MAM-RP2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Pro 307 (Mamu-A*01) & Ile 21 - Met 119 (B2M) (Accession # Q5TM76 (Mamu-A*01) & Q6V7J5 (B2M)).