Learn More
MSC ZEP Manual Dishwashing Detergent, Highly Concentrated, 1 gal, 4/Pack
Catalog No. 502819344
Encompass
This highly concentrated manual dishwashing detergent is formulated to emulsify tough grease and grime effectively. As part of the environmentally preferred GreenLink line, it is phosphate-free, fragrance-free, and features a neutral pH. Durable suds and water-softening agents ensure exceptional cleaning performance in both hard and soft water, providing a free-rinsing solution that leaves no hard water film.
  • Emulsifies tough grease and grime for spotless, residue-free results
  • Concentrated formula provides durable suds for economical, long-lasting cleaning
  • Water-softening agents ensure consistent performance in hard or soft water
  • Phosphate-free and eco-friendly, supporting environmentally preferable practices
  • Fragrance-free with a neutral pH formula
  • Clear green liquid appearance
Catalog No. 50-281-9344 Supplier MSC Supplier No. 17595067
This highly concentrated manual dishwashing detergent is formulated to emulsify tough grease and grime effectively. As part of the environmentally preferred GreenLink line, it is phosphate-free, fragrance-free, and features a neutral pH. Durable suds and water-softening agents ensure exceptional cleaning performance in both hard and soft water, providing a free-rinsing solution that leaves no hard water film.
  • Emulsifies tough grease and grime for spotless, residue-free results
  • Concentrated formula provides durable suds for economical, long-lasting cleaning
  • Water-softening agents ensure consistent performance in hard or soft water
  • Phosphate-free and eco-friendly, supporting environmentally preferable practices
  • Fragrance-free with a neutral pH formula
  • Clear green liquid appearance