Learn More
MSC ZEP Pot and Pan Detergent, Liquid, Lemon Scent, 5 gal
Catalog No. 502819111
Encompass
Clear Blue Pot and Pan Detergent is a liquid detergent formulated for manual dishwashing, supplied in a 5-gallon pail. It features a lemon scent and is designed for effective cleaning of kitchenware.
  • Liquid formulation for easy dispensing
  • Designed for manual dishwashing applications
  • Supplied in a 5 gal pail for bulk use
  • Features a characteristic lemon scent
  • Clear, blue viscous liquid appearance
  • Has a pH range of 7 to 8
  • Water soluble for easy rinsing
Catalog No. 50-281-9111 Supplier MSC Supplier No. 17595760
Clear Blue Pot and Pan Detergent is a liquid detergent formulated for manual dishwashing, supplied in a 5-gallon pail. It features a lemon scent and is designed for effective cleaning of kitchenware.
  • Liquid formulation for easy dispensing
  • Designed for manual dishwashing applications
  • Supplied in a 5 gal pail for bulk use
  • Features a characteristic lemon scent
  • Clear, blue viscous liquid appearance
  • Has a pH range of 7 to 8
  • Water soluble for easy rinsing