Learn More
Aviva Systems Biology Corp MAN1A2 Antibody - middle region (ARP46766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAN1A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAN1A2. Alternate names: MAN1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59394