Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mandelic acid-2,3,4,5,6-d5 | 70838-71-0 | 98.4% | 157.18 | C8H3D5O3 | 250 MG
SDP

Catalog No. 5000302571
Encompass_Preferred

Mandelic acid-2,3,4,5,6-d5 is a deuterium-labeled analogue of mandelic acid supplied as a white to off-white solid. It is intended for use as an isotopically labeled internal standard or tracer in analytical workflows such as LC-MS, supporting accurate quantitation, method validation, and tracer studies in research and bioanalysis.

  • Deuterium-labeled internal standard for LC-MS applications.
  • High purity (≈98.4%) for reliable quantitation.
  • Molecular formula C8H3D5O3; molecular weight 157.18 Da.
  • Supplied as a solid suitable for storage and precise weighing.
  • Available in small-scale sizes appropriate for analytical use.
  • Suitable for method development, validation, and tracer experiments.

Catalog No. 50-003-02571 Supplier Medchemexpress LLC Supplier No. 250MGHYW015591S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mandelic acid-2,3,4,5,6-d5 is a deuterium-labeled analogue of mandelic acid supplied as a white to off-white solid. It is intended for use as an isotopically labeled internal standard or tracer in analytical workflows such as LC-MS, supporting accurate quantitation, method validation, and tracer studies in research and bioanalysis.

  • Deuterium-labeled internal standard for LC-MS applications.
  • High purity (≈98.4%) for reliable quantitation.
  • Molecular formula C8H3D5O3; molecular weight 157.18 Da.
  • Supplied as a solid suitable for storage and precise weighing.
  • Available in small-scale sizes appropriate for analytical use.
  • Suitable for method development, validation, and tracer experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.