Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mandelic acid-2,3,4,5,6-d5 | 70838-71-0 | 98.4% | 157.18 g/mol | C8H3D5O3 | 50 MG
SDP

Catalog No. 5000301527
Encompass_Preferred

Mandelic acid-2,3,4,5,6-d5 is a deuterium-labeled mandelic acid used as an internal standard and tracer in analytical workflows. The isotopically labeled compound produces a defined mass shift for accurate quantification and is intended for research and analytical applications in mass spectrometry.

  • Deuterium-labeled at positions 2,3,4,5,6 (d5).
  • High purity (98.42%).
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Provided as a 50 mg analytical pack.
  • Molecular weight 157.18 g/mol; formula C8H3D5O3.
  • COA and SDS available for quality and safety verification.

Catalog No. 50-003-01527 Supplier Medchemexpress LLC Supplier No. HYW015591S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mandelic acid-2,3,4,5,6-d5 is a deuterium-labeled mandelic acid used as an internal standard and tracer in analytical workflows. The isotopically labeled compound produces a defined mass shift for accurate quantification and is intended for research and analytical applications in mass spectrometry.

  • Deuterium-labeled at positions 2,3,4,5,6 (d5).
  • High purity (98.42%).
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Provided as a 50 mg analytical pack.
  • Molecular weight 157.18 g/mol; formula C8H3D5O3.
  • COA and SDS available for quality and safety verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.