Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mandelic acid-2,3,4,5,6-d5 | 70838-71-0 | MFCD06658919 | 98.4% | 157.18 | C8H3D5O3 | 5 MG
SDP

Catalog No. 5000300728
Encompass_Preferred

Mandelic acid-2,3,4,5,6-d5 is a deuterated (D5) racemic analogue of mandelic acid intended for use as a stable-isotope internal standard and research reagent in analytical chemistry and tracer studies. It is characterized by high chemical purity and is provided in small to preparative quantities for method development, quantitation, and reference applications.

  • Deuterated (D5) analogue suitable for use as a stable-isotope internal standard.
  • High purity (98.42%) appropriate for analytical applications.
  • Molecular formula C8H3D5O3 and molecular weight 157.18 g·mol-1.
  • Supplied as a racemic (±) mixture for general analytical use.
  • Available in milligram to gram quantities for method development and reference standards.

Catalog No. 50-003-00728 Supplier Medchemexpress LLC Supplier No. HYW015591S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mandelic acid-2,3,4,5,6-d5 is a deuterated (D5) racemic analogue of mandelic acid intended for use as a stable-isotope internal standard and research reagent in analytical chemistry and tracer studies. It is characterized by high chemical purity and is provided in small to preparative quantities for method development, quantitation, and reference applications.

  • Deuterated (D5) analogue suitable for use as a stable-isotope internal standard.
  • High purity (98.42%) appropriate for analytical applications.
  • Molecular formula C8H3D5O3 and molecular weight 157.18 g·mol-1.
  • Supplied as a racemic (±) mixture for general analytical use.
  • Available in milligram to gram quantities for method development and reference standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.