Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Mandelonitrile benzoate | 95% | CAS 4242-46-0 | C15H11NO2 | 5g

Catalog No. 5001004226
Encompass_Preferred

Mandelonitrile benzoate | 95% | CAS 4242-46-0 | C15H11NO2 | 5g

Catalog No. 50-010-04226 Supplier Aobchem Supplier No. A6091725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mandelonitrile benzoate | 95% | CAS 4242-46-0 | C15H11NO2 | 5g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.