Learn More
Aviva Systems Biology Corp MANEA Antibody - middle region (ARP49845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MANEA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MANEA. Alternate names: DKFZp686D20120, FLJ12838, hEndo, ENDOApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53605