Learn More
Medchemexpress LLC Mangafodipir (trisodium) | 140678-14-4 | 99.7% | 1 ML
SDP
Catalog No. 5000407497
Encompass_Preferred
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent. It functions as an effective inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. This agent demonstrates no negative interference with the tumoricidal activity of chemotherapy.
  • Hepatocellular-specific contrast agent
  • Inhibits chemotherapy-induced peripheral neuropathy (CIPN)
  • Inhibits conditions caused by cellular oxidative stress
  • Shows no negative interference with chemotherapy's tumoricidal activity
  • Inhibits the effect of HGrC cell viability caused by H2O2
  • Attenuates apoptosis caused by cisplatin in HGrC cells
  • Shows protective effects against ovarian damage caused by cisplatin and paclitaxel in mice
Catalog No. 50-004-07497 Supplier Medchemexpress LLC Supplier No. HYB099310MM/1ML
Add to Cart
Add to Cart
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent. It functions as an effective inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. This agent demonstrates no negative interference with the tumoricidal activity of chemotherapy.
  • Hepatocellular-specific contrast agent
  • Inhibits chemotherapy-induced peripheral neuropathy (CIPN)
  • Inhibits conditions caused by cellular oxidative stress
  • Shows no negative interference with chemotherapy's tumoricidal activity
  • Inhibits the effect of HGrC cell viability caused by H2O2
  • Attenuates apoptosis caused by cisplatin in HGrC cells
  • Shows protective effects against ovarian damage caused by cisplatin and paclitaxel in mice