Learn More
Medchemexpress LLC Mangafodipir trisodium | 140678-14-4 | 99.7% | 100 MG
SDP
Catalog No. 5000407496
Encompass_Preferred
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent and an efficacious inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. It does not negatively interfere with the tumoricidal activity of chemotherapy.
  • Inhibits effect of HGrC cell viability caused by H2O2
  • Attenuates apoptosis caused by cisplatin in HGrC cells
  • Shows protective effects against ovarian damage caused by cisplatin and paclitaxel in mice
Catalog No. 50-004-07496 Supplier Medchemexpress LLC Supplier No. HYB0993100MG
Add to Cart
Add to Cart
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent and an efficacious inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. It does not negatively interfere with the tumoricidal activity of chemotherapy.
  • Inhibits effect of HGrC cell viability caused by H2O2
  • Attenuates apoptosis caused by cisplatin in HGrC cells
  • Shows protective effects against ovarian damage caused by cisplatin and paclitaxel in mice