Learn More
Medchemexpress LLC Mangafodipir (trisodium) | 140678-14-4 | 99.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0993
Encompass_Preferred
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent and an effective inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. It does not negatively interfere with the tumoricidal activity of chemotherapy.
  • Hepatocellular-specific contrast agent
  • Efficacious inhibitor of CIPN and other conditions caused by cellular oxidative stress
  • Shows no negative interference with the tumoricidal activity of chemotherapy
Catalog No. 50-004-50888