Learn More
Medchemexpress LLC Mangafodipir trisodium | 140678-14-4 | 99.7% | 5 MG
SDP
Catalog No. 5000407513
Encompass_Preferred
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent and an efficacious inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. It shows no negative interference with the tumoricidal activity of chemotherapy.
  • Hepatocellular-specific contrast agent
  • Efficacious inhibitor of chemotherapy-induced peripheral neuropathy
  • Attenuates apoptosis caused by cisplatin in HGrC cells
  • Protects against ovarian damage caused by cisplatin and paclitaxel in mice
Catalog No. 50-004-07513 Supplier Medchemexpress LLC Supplier No. HYB09935MG
Mangafodipir trisodium (MnDPDP) is a hepatocellular-specific contrast agent and an efficacious inhibitor of chemotherapy-induced peripheral neuropathy (CIPN) and other conditions caused by cellular oxidative stress. It shows no negative interference with the tumoricidal activity of chemotherapy.
  • Hepatocellular-specific contrast agent
  • Efficacious inhibitor of chemotherapy-induced peripheral neuropathy
  • Attenuates apoptosis caused by cisplatin in HGrC cells
  • Protects against ovarian damage caused by cisplatin and paclitaxel in mice